Assay Details
| Gene Symbol: LOC102190194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040746.1 | XP_017896235.1 | 4 - 5 | 842 | 73 | |
| XM_018040747.1 | XP_017896236.1 | 4 - 5 | 842 | 73 | ||
| XM_018040748.1 | XP_017896237.1 | 4 - 5 | 842 | 73 | ||
| XM_018040749.1 | XP_017896238.1 | 2 - 3 | 604 | 73 | ||
| GenBank mRNA | JT752588.1 | - | 5 - 6 | 404 | 73 | |
More Information
Set Membership: |
|

LOC102190194
Genomic Map