Assay Details
Gene Symbol: LOC102183580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700450.3 | XP_005700507.2 | 1 | 668 | 93 | |
GenBank mRNA | JO648987.2 | - | 1 | 426 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
