Assay Details
| Gene Symbol: LOC102183049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005702103.3 | XP_005702160.1 | 1 | 2288 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102183049| Gene Symbol: LOC102183049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005702103.3 | XP_005702160.1 | 1 | 2288 | 72 | |
Set Membership: |
|