Assay Details
| Gene Symbol: RP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700814.3 | XP_005700871.1 | 1 - 2 | 300 | 92 | |
| GenBank mRNA | JO440934.2 | - | 2 - 3 | 752 | 92 | |
More Information
Set Membership: |
|

RP2
Genomic Map