Assay Details
| Gene Symbol: LOC102184794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045476.1 | XP_017900965.1 | 3494 | 77 | ||
| GenBank mRNA | JO635087.2 | - | 3567 | 77 | ||
| KA326520.1 | - | 3501 | 77 | |||
| Gene Symbol: LOC108633277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048960.1 | XP_017904449.1 | 3063 | 77 | ||
| GenBank mRNA | JO422084.2 | - | 3063 | 77 | ||
| JO426161.1 | - | 489 | 77 | |||
| Gene Symbol: LOC108636083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048969.1 | XP_017904458.1 | 2 - 3 | 324 | 77 | |
More Information
Set Membership: |
|

LOC102184794
Genomic Map