Assay Details
| Gene Symbol: NDUFA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050998.1 | XP_017906487.1 | 3 - 4 | 305 | 94 | |
| GenBank mRNA | JO459015.2 | - | 3 - 4 | 298 | 94 | |
| JO643044.2 | - | 1 - 2 | 218 | 94 | ||
| KA334170.1 | - | 3 - 4 | 280 | 94 | ||
More Information
Set Membership: |
|

NDUFA7
Genomic Map