Assay Details
| Gene Symbol: TIPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053621.1 | XP_017909110.1 | 3 - 4 | 312 | 75 | |
| XM_018053622.1 | XP_017909111.1 | 4 - 5 | 322 | 75 | ||
| XM_018053623.1 | XP_017909112.1 | 3 - 4 | 389 | 75 | ||
| XM_018053624.1 | XP_017909113.1 | 2 - 3 | 162 | 75 | ||
| XM_018053625.1 | XP_017909114.1 | 1 - 2 | 305 | 75 | ||
| GenBank mRNA | JO339935.2 | - | 1 - 2 | 85 | 75 | |
| KA333207.1 | - | 3 - 4 | 293 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018053624.1, XM_018053625.1 transcripts.
Set Membership: |
|

TIPIN
Genomic Map