Assay Details
| Gene Symbol: LOC102187872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044794.1 | XP_017900283.1 | 2185 | 59 | ||
| GenBank mRNA | JO373579.2 | - | 298 | 59 | ||
| Gene Symbol: COL5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056198.1 | XP_017911687.1 | 2187 | 59 | ||
| GenBank mRNA | JO538678.2 | - | 2457 | 59 | ||
| KA342935.1 | - | 3908 | 59 | |||
More Information
Set Membership: |
|

LOC102187872
Genomic Map