Assay Details
| Gene Symbol: LOC102187872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044794.1 | XP_017900283.1 | 215 | 58 | ||
| GenBank mRNA | JO373579.2 | - | 2268 | 58 | ||
| JO422347.2 | - | 1690 | 58 | |||
| Gene Symbol: COL5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056198.1 | XP_017911687.1 | 217 | 58 | ||
| GenBank mRNA | JO538678.2 | - | 487 | 58 | ||
| KA342935.1 | - | 5878 | 58 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018056198.1 transcripts.
Set Membership: |
|

LOC102187872
Genomic Map