Assay Details
| Gene Symbol: LOC102187872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044794.1 | XP_017900283.1 | 1277 | 116 | ||
| GenBank mRNA | JO373579.2 | - | 1206 | 116 | ||
| JO422347.2 | - | 628 | 116 | |||
| Gene Symbol: COL5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056198.1 | XP_017911687.1 | 1279 | 116 | ||
| GenBank mRNA | JO538678.2 | - | 1549 | 116 | ||
| KA342935.1 | - | 4816 | 116 | |||
More Information
Set Membership: |
|

LOC102187872
Genomic Map