Assay Details
| Gene Symbol: LOC106502686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056180.1 | XP_017911669.1 | 2 - 3 | 405 | 69 | |
| GenBank mRNA | JO400142.2 | - | 2 - 3 | 264 | 69 | |
More Information
Set Membership: |
|

LOC106502686
Genomic Map