Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061866.1 | XP_017917355.1 | 8 - 9 | 1343 | 61 | |
| XM_018061868.1 | XP_017917357.1 | 7 - 8 | 1265 | 61 | ||
| XM_018061869.1 | XP_017917358.1 | 8 - 9 | 1122 | 61 | ||
| XM_018061870.1 | XP_017917359.1 | 6 - 7 | 972 | 61 | ||
| XM_018061871.1 | XP_017917360.1 | 8 - 9 | 1368 | 61 | ||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map