Assay Details
| Gene Symbol: MUSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067100.1 | XP_017922589.1 | 2 - 3 | 938 | 82 | |
| GenBank mRNA | KA318594.1 | - | 2 - 3 | 351 | 82 | |
More Information
Set Membership: |
|

MUSTN1
Genomic Map