Assay Details
| Gene Symbol: FGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056811.1 | XP_017912300.1 | 9 - 10 | 1303 | 63 | |
| XM_018056817.1 | XP_017912306.1 | 9 - 10 | 1314 | 63 | ||
| XM_018056824.1 | XP_017912313.1 | 9 - 10 | 1300 | 63 | ||
| XM_018056826.1 | XP_017912315.1 | 9 - 10 | 1312 | 63 | ||
| GenBank mRNA | JO339531.2 | - | 10 - 11 | 1390 | 63 | |
| JO580069.1 | - | 4 - 5 | 1783 | 63 | ||
More Information
Set Membership: |
|

FGR
Genomic Map