Assay Details
| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040796.1 | XP_017896285.1 | 9 - 10 | 1450 | 65 | |
| GenBank mRNA | KA307936.1 | - | 9 - 10 | 1419 | 65 | |
More Information
Set Membership: |
|

BICDL2
Genomic Map