Assay Details
| Gene Symbol: LIPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975084.2 | XP_013830538.2 | 5 - 6 | 711 | 84 | |
| XM_018041737.1 | XP_017897226.1 | 6 - 7 | 785 | 84 | ||
| XM_018041738.1 | XP_017897227.1 | 6 - 7 | 782 | 84 | ||
| XM_018041739.1 | XP_017897228.1 | 5 - 6 | 722 | 84 | ||
| XM_018041740.1 | XP_017897229.1 | 6 - 7 | 772 | 84 | ||
| XM_018041741.1 | XP_017897230.1 | 6 - 7 | 769 | 84 | ||
More Information
Set Membership: |
|

LIPN
Genomic Map