Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675700.3 | XP_005675757.2 | 1 - 2 | 130 | 96 | |
| Gene Symbol: APCDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO411231.2 | - | 7 - 8 | 983 | 96 | |
More Information
Set Membership: |
|

VPS26C
Genomic Map