Assay Details
| Gene Symbol: CSDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681114.3 | XP_005681171.1 | 3 - 4 | 767 | 61 | |
| GenBank mRNA | JO632487.2 | - | 3 - 4 | 570 | 61 | |
| KA323684.1 | - | 3 - 4 | 569 | 61 | ||
More Information
Set Membership: |
|

CSDC2
Genomic Map