Assay Details
| Gene Symbol: QDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049755.1 | XP_017905244.1 | 5 - 6 | 589 | 72 | |
| GenBank mRNA | KA338431.1 | - | 5 - 6 | 600 | 72 | |
More Information
Set Membership: |
|

QDPR
Genomic Map