Assay Details
| Gene Symbol: SAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682944.3 | XP_005683001.2 | 3 - 4 | 208 | 71 | |
| GenBank mRNA | JT635369.1 | - | 5 - 6 | 617 | 71 | |
More Information
Set Membership: |
|

SAR1B
Genomic Map