Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683172.3 | XP_005683229.1 | 2 - 3 | 412 | 62 | |
| XM_005683173.3 | XP_005683230.1 | 2 - 3 | 393 | 62 | ||
| GenBank mRNA | JO580974.1 | - | 2 - 3 | 457 | 62 | |
More Information
Set Membership: |
|

RPS14
Genomic Map