Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054922.1 | XP_017910411.1 | 5 - 6 | 777 | 62 | |
| GenBank mRNA | JO621954.2 | - | 2 - 3 | 1976 | 62 | |
More Information
Set Membership: |
|

CHST10
Genomic Map