Assay Details
| Gene Symbol: LIMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686647.3 | XP_005686704.1 | 4 - 5 | 718 | 74 | |
| XM_018055308.1 | XP_017910797.1 | 4 - 5 | 822 | 74 | ||
| XM_018055309.1 | XP_017910798.1 | 4 - 5 | 674 | 74 | ||
| XM_018055310.1 | XP_017910799.1 | 4 - 5 | 502 | 74 | ||
| XM_018055311.1 | XP_017910800.1 | 4 - 5 | 497 | 74 | ||
| XM_018055312.1 | XP_017910801.1 | 4 - 5 | 1125 | 74 | ||
More Information
Set Membership: |
|

LIMS1
Genomic Map