Assay Details
| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055580.1 | XP_017911069.1 | 6 - 7 | 1314 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM228A| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055580.1 | XP_017911069.1 | 6 - 7 | 1314 | 66 | |
Set Membership: |
|