Assay Details
| Gene Symbol: ARHGAP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057103.1 | XP_017912592.1 | 13 - 14 | 3411 | 59 | |
| XM_018057104.1 | XP_017912593.1 | 14 - 15 | 3568 | 59 | ||
| XM_018057105.1 | XP_017912594.1 | 13 - 14 | 3234 | 59 | ||
| XM_018057106.1 | XP_017912595.1 | 12 - 13 | 3381 | 59 | ||
| GenBank mRNA | JT741711.1 | - | 18 - 19 | 3569 | 59 | |
More Information
Set Membership: |
|

ARHGAP21
Genomic Map