Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691434.3 | XP_005691491.1 | 4 - 5 | 777 | 84 | |
| XM_005691435.2 | XP_005691492.1 | 4 - 5 | 465 | 84 | ||
| GenBank mRNA | JO618834.2 | - | 2 - 3 | 252 | 84 | |
| JT723162.1 | - | 2 - 3 | 205 | 84 | ||
More Information
Set Membership: |
|

FAM216A
Genomic Map