Assay Details
| Gene Symbol: SPG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061889.1 | XP_017917378.1 | 11 - 12 | 1590 | 61 | |
| XM_018061890.1 | XP_017917379.1 | 11 - 12 | 1580 | 61 | ||
More Information
Set Membership: |
|

SPG7
Genomic Map