Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692363.3 | XP_005692420.1 | 3 - 4 | 1075 | 86 | |
| GenBank mRNA | JT702739.1 | - | 2 - 3 | 180 | 86 | |
| KA312284.1 | - | 2 - 3 | 245 | 86 | ||
More Information
Set Membership: |
|

ZNF570
Genomic Map