Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692363.3 | XP_005692420.1 | 4 - 5 | 1162 | 95 | |
| GenBank mRNA | KA312284.1 | - | 1 - 2 | 158 | 95 | |
More Information
Set Membership: |
|

ZNF570
Genomic Map