Assay Details
| Gene Symbol: RSPH6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692787.3 | XP_005692844.2 | 3 - 4 | 1789 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPH6A| Gene Symbol: RSPH6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692787.3 | XP_005692844.2 | 3 - 4 | 1789 | 53 | |
Set Membership: |
|