Assay Details
Gene Symbol: KRT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693792.2 | XP_005693849.2 | 3 - 4 | 804 | 53 | |
XM_018065056.1 | XP_017920545.1 | 3 - 4 | 798 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
