Assay Details
| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695059.3 | XP_005695116.2 | 2 - 3 | 1180 | 99 | |
| GenBank mRNA | JO331375.2 | - | 1 - 2 | 285 | 99 | |
| JO581743.1 | - | 1 - 2 | 660 | 99 | ||
More Information
Set Membership: |
|

BCL2A1
Genomic Map