Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695506.3 | XP_005695563.2 | 4 - 5 | 930 | 64 | |
| XM_013973395.2 | XP_013828849.2 | 4 - 5 | 930 | 64 | ||
| XM_018067029.1 | XP_017922518.1 | 5 - 6 | 923 | 64 | ||
| XM_018067030.1 | XP_017922519.1 | 5 - 6 | 928 | 64 | ||
| GenBank mRNA | JO612210.2 | - | 5 - 6 | 2197 | 64 | |
More Information
Set Membership: |
|

AZI2
Genomic Map