Assay Details
| Gene Symbol: SRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696330.3 | XP_005696387.2 | 6 - 7 | 1744 | 56 | |
| GenBank mRNA | JO595619.2 | - | 6 - 7 | 1100 | 56 | |
| JO640878.2 | - | 6 - 7 | 1495 | 56 | ||
More Information
Set Membership: |
|

SRF
Genomic Map