Assay Details
| Gene Symbol: LOC102174532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697888.3 | XP_005697945.1 | 16 - 17 | 2247 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102174532| Gene Symbol: LOC102174532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697888.3 | XP_005697945.1 | 16 - 17 | 2247 | 101 | |
Set Membership: |
|