Assay Details
| Gene Symbol: PNLIPRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698515.3 | XP_005698572.1 | 9 - 10 | 1076 | 56 | |
| XM_013975301.2 | XP_013830755.1 | 8 - 9 | 1114 | 56 | ||
| XM_013975302.2 | XP_013830756.1 | 7 - 8 | 941 | 56 | ||
More Information
Set Membership: |
|

PNLIPRP2
Genomic Map