Assay Details
| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005701104.3 | XP_005701161.1 | 3 - 4 | 382 | 65 | |
| GenBank mRNA | JO647257.2 | - | 2 - 3 | 178 | 65 | |
More Information
Set Membership: |
|

TRAPPC2
Genomic Map