Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047491.1 | XP_017902980.1 | 7 - 8 | 1949 | 65 | |
| GenBank mRNA | JO424658.1 | - | 5 - 6 | 1167 | 65 | |
More Information
Set Membership: |
|

PODXL
Genomic Map