Assay Details
| Gene Symbol: ZC3HAV1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046886.1 | XP_017902375.1 | 3 - 4 | 1101 | 71 | |
| GenBank mRNA | KA350581.1 | - | 1 - 2 | 754 | 71 | |
More Information
Set Membership: |
|

ZC3HAV1L
Genomic Map