Assay Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047730.1 | XP_017903219.1 | 5 - 6 | 2125 | 72 | |
| XM_018047731.1 | XP_017903220.1 | 5 - 6 | 2056 | 72 | ||
| XM_018047732.1 | XP_017903221.1 | 6 - 7 | 2068 | 72 | ||
| XM_018047733.1 | XP_017903222.1 | 6 - 7 | 1807 | 72 | ||
| XM_018047734.1 | XP_017903223.1 | 6 - 7 | 1738 | 72 | ||
More Information
Set Membership: |
|

NR4A1
Genomic Map