Assay Details
| Gene Symbol: MARCH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048003.1 | XP_017903492.1 | 3 - 4 | 904 | 64 | |
| GenBank mRNA | JO432032.2 | - | 1 - 2 | 870 | 64 | |
| JT757360.1 | - | 5 - 6 | 995 | 64 | ||
More Information
Set Membership: |
|

MARCH9
Genomic Map