Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964573.2 | XP_013820027.2 | 2 - 3 | 4789 | 65 | |
| XM_018049355.1 | XP_017904844.1 | 3 - 4 | 4687 | 65 | ||
More Information
Set Membership: |
|

PCDH7
Genomic Map