Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050386.1 | XP_017905875.1 | 2 - 3 | 692 | 81 | |
| GenBank mRNA | JO579383.1 | - | 5 - 6 | 1596 | 81 | |
More Information
Set Membership: |
|

PPP2CA
Genomic Map