Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053697.1 | XP_017909186.1 | 11 - 12 | 2148 | 57 | |
| GenBank mRNA | JO639389.2 | - | 3 - 4 | 419 | 57 | |
More Information
Set Membership: |
|

SV2C
Genomic Map