Assay Details
| Gene Symbol: AMACR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065752.1 | XP_017921241.1 | 2 - 3 | 482 | 67 | |
| GenBank mRNA | JO434445.2 | - | 2 - 3 | 516 | 67 | |
More Information
Set Membership: |
|

AMACR
Genomic Map