Assay Details
| Gene Symbol: RAB44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973756.2 | XP_013829210.2 | 9 - 10 | 2579 | 55 | |
| GenBank mRNA | KA343400.1 | - | 1 - 2 | 237 | 55 | |
More Information
Set Membership: |
|

RAB44
Genomic Map