Assay Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039206.1 | XP_017894695.1 | 2 | 208 | 86 | |
| GenBank mRNA | JT728211.1 | - | 6 | 935 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM13| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039206.1 | XP_017894695.1 | 2 | 208 | 86 | |
| GenBank mRNA | JT728211.1 | - | 6 | 935 | 86 | |
Set Membership: |
|