Assay Details
| Gene Symbol: LOC102181879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045406.1 | XP_017900895.1 | 2376 | 60 | ||
| Gene Symbol: DSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038784.1 | XP_017894273.1 | 11 - 12 | 1689 | 60 | |
| XM_018038785.1 | XP_017894274.1 | 11 - 12 | 1568 | 60 | ||
More Information
Set Membership: |
|

LOC102181879
Genomic Map