Assay Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040395.1 | XP_017895884.1 | 11 - 12 | 1344 | 55 | |
| XM_018040396.1 | XP_017895885.1 | 13 - 14 | 1810 | 55 | ||
| GenBank mRNA | JO441844.2 | - | 2 - 3 | 371 | 55 | |
More Information
Set Membership: |
|

LRWD1
Genomic Map