Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065260.1 | XP_017920749.1 | 10 - 11 | 1648 | 64 | |
| XM_018065263.1 | XP_017920752.1 | 9 - 10 | 1609 | 64 | ||
| XM_018065272.1 | XP_017920761.1 | 7 - 8 | 1139 | 64 | ||
More Information
Set Membership: |
|

ZNF385B
Genomic Map